missing translation for 'onlineSavingsMsg'
Learn More

11-Azido-3,6,9-trioxaundecan-1-amine 95.0+%, TCI America™

Catalog No. A23635G
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23635G 5 g
A2363200MG 200 mg
A23631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A23635G Supplier TCI America Supplier No. A23635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134179-38-7
Molecular Formula C8H18N4O3
Molecular Weight (g/mol) 218.257
MDL Number MFCD00269874
InChI Key FPVCVHVTMPCZTH-UHFFFAOYSA-N
Synonym 1-amino-11-azido-3,6,9-trioxaundecane, azido-peg3-amine, 11-azido-3,6,9-trioxaundecan-1-amine, 2-2-2-2-azidoethoxy ethoxy ethoxy ethanamine, 1-2-2-aminoethoxy ethoxy-2-2-azidoethoxy ethane, 11-azido-3 6 9-trioxaundecan-1-amine, ethanamine, 2-2-2-2-azidoethoxy ethoxy ethoxy, 2-2-2-2-azidoethoxy ethoxy ethoxy ethylamine
PubChem CID 2735280
IUPAC Name 2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethanamine
SMILES C(COCCOCCOCCN=[N+]=[N-])N

Specifications

Quantity 5 g
UN Number 2735
Formula Weight 218.26
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 11-Azido-3,6,9-trioxaundecan-1-amine
TSCA No
Recommended Storage Refrigerator