missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Reaction Buffer with MgCl2 for DNase I (10X)
Description
10X Reaction Buffer with MgCl2 is used with DNase I, RNase-free.
Specifications
Specifications
| Concentration | 10X |
| Quantity | 1 mL |
| Product Type | Reaction Buffer |
| Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.