missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 10058-F4

Numéro de catalogue. 440610
Modifier l'affichage
Click to view available options
Quantité:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
440610 10 mg
440650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 440610 Fournisseur Tocris Bioscience™ Code fournisseur 4406/10

Inhibits c-Myc-Max dimerization

10058-F4 is a cell permeable c-Myc-Max dimerization inhibitor. Inhibits proliferation, induces apoptosis and arrests cells in G0/G1 in rat1a-c-Myc cells. Also reduces tumor growth in vivo. In lymphoma cells, 10058-F4 decreases glucose uptake and expression of glycolysis-associated genes and reduces cell viability.

Identifiants chimiques

CAS 403811-55-2
Formule moléculaire C12H11NOS2

Spécifications

Nom ou substance chimique 10058-F4
Quantité 10 mg
Poids de formule 249.35
Forme physique Solid
Couleur Yellow
Pureté >99%
Poids moléculaire (g/mol) 249.346
Recommended Storage Store at +4°C