1000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.
Numéro de catalogue.
509558955
Fournisseur
Ace GlassCode fournisseur
1203519
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
1000mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 4 cord with locking connector. cULus rated. CSA certified.