missing translation for 'onlineSavingsMsg'
Learn More

10-Methylacridinium Perchlorate 98.0+%, TCI America™

Catalog No. M17871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17871G Supplier TCI America Supplier No. M17871G
Only null left

Chemical Identifiers

CAS 26456-05-3
Molecular Formula C14H12ClNO4
Molecular Weight (g/mol) 293.703
InChI Key UTTIEKPPJSQKJC-UHFFFAOYSA-M
PubChem CID 11808515
IUPAC Name 10-methylacridin-10-ium;perchlorate
SMILES C[N+]1=C2C=CC=CC2=CC3=CC=CC=C31.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 241°C
Quantity 1 g
UN Number 1479
Formula Weight 293.70
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 10-Methylacridinium Perchlorate