missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxydecanoic acid, 96%

Catalog No. AAH6074306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6074306 5 g
AAH6074303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6074306 Supplier Thermo Scientific Chemicals Supplier No. H6074306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1679-53-4 | C10H20O3 | 188.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1679-53-4
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.27
MDL Number MFCD00010510
InChI Key YJCJVMMDTBEITC-UHFFFAOYSA-N
Synonym 10-hydroxydecanoate, decanoic acid, 10-hydroxy, unii-np03xo416b, 10-hydroxy capric acid, 10-hydroxy-decanoic acid, 1-hydroxydecanoicacid, 10-oh-capric acid, 10-hydroxycapric acid, 10-oh-decanoic acid, 10-hydroxydecanoicacid
PubChem CID 74300
ChEBI CHEBI:17409
IUPAC Name 10-hydroxydecanoic acid
SMILES OCCCCCCCCCC(O)=O

Specifications

Melting Point 75°C to 77°C
Linear Formula HO(CH2)9CO2H
Quantity 5 g
Formula Weight 188.26
Percent Purity 96%
Chemical Name or Material 10-Hydroxydecanoic acid
EINECSNumber 216-848-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only