missing translation for 'onlineSavingsMsg'
Learn More

10-Bromodecanoic acid, 95%

Catalog No. AAH6029214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6029214 25 g
AAH6029206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6029214 Supplier Thermo Scientific Chemicals Supplier No. H6029214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 50530-12-6 | C10H19BrO2 | 251.164 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 50530-12-6
Molecular Formula C10H19BrO2
Molecular Weight (g/mol) 251.164
MDL Number MFCD00014388
InChI Key PGVRSPIEZYGOAD-UHFFFAOYSA-N
Synonym 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
PubChem CID 142712
IUPAC Name 10-bromodecanoic acid
SMILES C(CCCCC(=O)O)CCCCBr

Specifications

Melting Point 38°C to 41°C
Linear Formula Br(CH2)9CO2H
Quantity 25 g
Solubility Information Partly miscible in water.
Formula Weight 251.16
Percent Purity 95%
Chemical Name or Material 10-Bromodecanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only