missing translation for 'onlineSavingsMsg'
Learn More

10-Bromo-1-decanol, 95%

Catalog No. AAB2148306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2148306 5 g
AAB2148303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2148306 Supplier Thermo Scientific Chemicals Supplier No. B2148306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53463-68-6 | C10H21BrO | 237.18 g/mol

10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10-Bromo-1-decanol is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 53463-68-6
Molecular Formula C10H21BrO
Molecular Weight (g/mol) 237.18
MDL Number MFCD00041681
InChI Key LGZMUUBPTDRQQM-UHFFFAOYSA-N
Synonym 10-bromo-1-decanol, 10-bromodecanol, 1-decanol, 10-bromo, 10-hydroxy-1-bromo decane, 1-bromodecan-10-ol, 10-bromodecane-1-ol, 10-bromo-decan-1-ol, decamethylene bromohydrin, 10-hydroxy1-bromo decane, acmc-209l7p
PubChem CID 104507
IUPAC Name 10-bromodecan-1-ol
SMILES OCCCCCCCCCCBr

Specifications

Density 1.19
Boiling Point 165°C to 169°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.476
Quantity 5 g
Beilstein 1738593
Solubility Information It is miscible with water.
Formula Weight 237.19
Percent Purity 95%
Chemical Name or Material 10-Bromo-1-decanol
Hazard Category H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-572-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only