missing translation for 'onlineSavingsMsg'
Learn More

10,10'-Dimethyl-9,9'-biacridinium Dinitrate 97.0+%, TCI America™

Catalog No. p-7118918
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12031G 1 g
B12035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12031G Supplier TCI America Supplier No. B12031G

Chemical Identifiers

CAS 2315-97-1
Molecular Formula C28H22N4O6
Molecular Weight (g/mol) 510.51
MDL Number MFCD00011925
InChI Key KNJDBYZZKAZQNG-UHFFFAOYSA-N
Synonym Lucigenin, Bis(N-methylacridinium) Nitrate
PubChem CID 129893650
IUPAC Name 10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium dinitrate
SMILES [O-][N+]([O-])=O.[O-][N+]([O-])=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12

Specifications

Melting Point 247°C
Quantity 1 g
UN Number 1479
Formula Weight 510.51
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 10,10′-Dimethyl-9,9′-biacridinium Dinitrate [for Chemiluminescence Research]