missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. V01455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V01455G Supplier TCI America Supplier No. V01455G

Chemical Identifiers

CAS 1013027-27-4
Molecular Formula C7H7F6N3O4S2
Molecular Weight (g/mol) 375.26
InChI Key OVFZHDDKAVGFIS-UHFFFAOYSA-N
PubChem CID 134128844
IUPAC Name 1-ethenylimidazole;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C=CN1C=CN=C1.C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 37°C
Quantity 5 g
Formula Weight 375.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Vinylimidazolium Bis(trifluoromethanesulfonyl)imide