missing translation for 'onlineSavingsMsg'
Learn More

1-Vinyl-2-pyrrolidone (stabilized with N,N'-Di-sec-butyl-p-phenylenediamine) 99.0+%, TCI America™

Catalog No. V0026500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0026500ML 500 mL
V002625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0026500ML Supplier TCI America Supplier No. V0026500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-12-0
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD00003197
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES C=CN1CCCC1=O

Specifications

Boiling Point 148°C
Quantity 500 mL
UN Number 2810
Formula Weight 111.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Vinyl-2-pyrrolidone (stabilized with N,N′-Di-sec-butyl-p-phenylenediamine)
EINECSNumber (5)-0114
RTECSNumber UY6107000
TSCA Yes
Recommended Storage Refrigerator