missing translation for 'onlineSavingsMsg'
Learn More

1-(Triisopropylsilyl)pyrrole 96.0+%, TCI America™

Catalog No. T16775ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16775ML 5 mL
T167725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16775ML Supplier TCI America Supplier No. T16775ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87630-35-1
Molecular Formula C13H25NSi
Molecular Weight (g/mol) 223.44
MDL Number MFCD00054932
InChI Key FBQURXLBJJNDBX-UHFFFAOYSA-N
Synonym 1-triisopropylsilyl pyrrole, 1-triisopropylsilyl-1h-pyrrole, n-triisopropylsilylpyrrole, 1h-pyrrole, 1-tris 1-methylethyl silyl, tri propan-2-yl-pyrrol-1-ylsilane, 1-tris propan-2-yl silyl-1h-pyrrole, pubchem9194, 1-triisopropylsilylpyrrole, 1-triisoproylsilyl-pyrrole, acmc-1bkd7
PubChem CID 145136
IUPAC Name 1-[tris(propan-2-yl)silyl]-1H-pyrrole
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 223.44
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Triisopropylsilyl)pyrrole