missing translation for 'onlineSavingsMsg'
Learn More

1-(Triisopropylsilyl)pyrrole, 95%

Catalog No. AAL1315214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1315214 25 g
AAL1315206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1315214 Supplier Thermo Scientific Chemicals Supplier No. L1315214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87630-35-1 | C13H25NSi | 223.44 g/mol

1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2?-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Triisopropylsilyl)pyrrole may be employed as reagent in perfluoroalkylation and Vilsmeier formylation reactions. It may be used in the preparation of ethyl 2-(2,4-dinitrophenylhydrazono]-3-[ 1-(triisopropylsily1)-pyrrol-2-yflpropanoate; heterocyclic base, 3-nitropyrrole and 3-nitropyrrole, required for the synthesis of 1 -(2′-deoxy-β-D-ribofuranosyl)-3-nitropyrrole. It participates in various electrophilic substitution reactions specifically at β-position, via reaction with various electrophilic reagents (Br+, I+,NO2+,etc).

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 87630-35-1
Molecular Formula C13H25NSi
Molecular Weight (g/mol) 223.44
MDL Number MFCD00054932
InChI Key FBQURXLBJJNDBX-UHFFFAOYSA-N
Synonym 1-triisopropylsilyl pyrrole, 1-triisopropylsilyl-1h-pyrrole, n-triisopropylsilylpyrrole, 1h-pyrrole, 1-tris 1-methylethyl silyl, tri propan-2-yl-pyrrol-1-ylsilane, 1-tris propan-2-yl silyl-1h-pyrrole, pubchem9194, 1-triisopropylsilylpyrrole, 1-triisoproylsilyl-pyrrole, acmc-1bkd7
PubChem CID 145136
IUPAC Name tri(propan-2-yl)-pyrrol-1-ylsilane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC=C1

Specifications

Density 0.904
Boiling Point 78°C (0.4 mmHg)
Flash Point 106°C (222°F)
Refractive Index 1.492
Quantity 25 g
Beilstein 3540663
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 223.44
Percent Purity 95%
Chemical Name or Material 1-(Triisopropylsilyl)pyrrole
Show More Show Less
Hazard Category H302+H332-H319
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only