missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 170°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 428.39 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one |
Safety and Handling
| TSCA | No |