missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%

Catalog No. AAH26055MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26055MD 250 mg
AAH2605503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26055MD Supplier Thermo Scientific Chemicals Supplier No. H26055MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 277756-44-2 | C7H9F3O2 | 182.142 g/mol

1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 277756-44-2
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.142
MDL Number MFCD08445821
InChI Key DGQRYPPBAJNZFZ-UHFFFAOYSA-N
Synonym 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl
PubChem CID 15507525
IUPAC Name 1-(trifluoromethyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)C(F)(F)F

Specifications

Melting Point 36°C to 38°C
Quantity 250 mg
Solubility Information Insoluble in water.
Formula Weight 182.15
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)cyclopentanecarboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only