missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethanesulfonyl)-1H-benzotriazole 98.0+%, TCI America™

Numéro de catalogue. T31671G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T31671G 1 g
T31675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T31671G Fournisseur TCI America Code fournisseur T31671G
Il en reste null

Identifiants chimiques

CAS 117632-84-5
Molecular Formula C7H4F3N3O2S
Molecular Weight (g/mol) 251.18
MDL Number MFCD00963686
InChI Key QTTZLMGXPHZYKR-UHFFFAOYSA-N
PubChem CID 3692410
IUPAC Name 1-trifluoromethanesulfonyl-1H-1,2,3-benzotriazole
SMILES FC(F)(F)S(=O)(=O)N1N=NC2=CC=CC=C12

Spécifications

Melting Point 38°C
Quantity 1 g
Formula Weight 251.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Trifluoromethanesulfonyl)-1H-benzotriazole
TSCA No
Recommended Storage Refrigerator