missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecyne, 97%

Catalog No. AAL0248106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0248106 5 g
AAL0248103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0248106 Supplier Thermo Scientific Chemicals Supplier No. L0248106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26186-02-7 | C13H24 | 180.34 g/mol

It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.

Chemical Identifiers

CAS 26186-02-7
Molecular Formula C13H24
Molecular Weight (g/mol) 180.34
MDL Number MFCD00015074
InChI Key GZEDKDBFUBPZNG-UHFFFAOYSA-N
Synonym 1-tridecyne, tridecyne, acmc-1cinm
PubChem CID 117754
IUPAC Name tridec-1-yne
SMILES CCCCCCCCCCCC#C

Specifications

Density 0.78
Boiling Point 116°C to 118°C (10 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.438
Linear Formula CH3(CH2)10C≡CH
Quantity 5 g
Beilstein 1744658
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 180.34
Percent Purity 97%
Chemical Name or Material 1-Tridecyne
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 247-511-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only