missing translation for 'onlineSavingsMsg'
Learn More

1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole 98.0+%, TCI America™

Catalog No. T34085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34085G 5 g
T34081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34085G Supplier TCI America Supplier No. T34085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 449758-17-2
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD11044692
InChI Key IMZWSOSYNFVECD-UHFFFAOYSA-N
Synonym 1-2-tetrahydropyranyl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 1-oxan-2-yl-1h-pyrazole, 1-oxan-2-yl pyrazole, 1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl, 1-2-tetrahydropyranyl pyrazole, 1-tetrahydro-pyran-2-yl-1h-pyrazole, 1-2-oxanyl pyrazole, ksc588k5l, 1-tetrahydropyran-2-ylpyrazole
PubChem CID 21866310
IUPAC Name 1-(oxan-2-yl)pyrazole
SMILES C1CCOC(C1)N2C=CC=N2

Specifications

Color Yellow
Boiling Point 65°C
Quantity 5 g
Formula Weight 152.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole