missing translation for 'onlineSavingsMsg'
Learn More

1-Tetracosanol 98.0+%, TCI America™

Catalog No. T05935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05935G 5 g
T05931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05935G Supplier TCI America Supplier No. T05935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 506-51-4
Molecular Formula C24H50O
Molecular Weight (g/mol) 354.66
MDL Number MFCD00042662
InChI Key TYWMIZZBOVGFOV-UHFFFAOYSA-N
Synonym 1-tetracosanol, tetracosanol, lignoceryl alcohol, lignocerol, n-tetracosanol, tetracosyl alcohol, lignoceric alcohol, unii-2n0pi37ioc, n-tetracosanol-1, 2n0pi37ioc
PubChem CID 10472
ChEBI CHEBI:77413
IUPAC Name tetracosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 354.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Tetracosanol
EINECSNumber (2)-0217
TSCA Yes