missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. CD07394CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CD07394CB 250mg
CD07394DA 1g
CD07394DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CD07394CB Fournisseur Thermo Scientific Chemicals Code fournisseur CD07394CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 647824-51-9
Formule moléculaire C10H16N2O
Poids moléculaire (g/mol) 180.251
Numéro MDL MFCD00120293
Clé InChI WQHXOHHEPNBXMF-UHFFFAOYSA-N
Synonyme 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c
PubChem CID 2776936
Nom de l’IUPAC 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde
SOURIRES CC1=C(C(=NN1C(C)(C)C)C)C=O

Spécifications

Nom ou substance chimique 1-(Tert-butyl)-3, 5-dimethyl-1H-pyrazole-4-carbaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 180.25
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.