missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-3-hydroxypiperidine 98.0+%, TCI America™

Catalog No. B32781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32781G Supplier TCI America Supplier No. B32781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85275-45-2
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD02093938
InChI Key UIJXHKXIOCDSEB-UHFFFAOYSA-N
Synonym 1-boc-3-hydroxypiperidine, 1-n-boc-3-hydroxy-piperidine, 1-boc-3-hydroxy-piperidine, n-boc-3-piperidinol, n-boc-3-hydroxypiperidine, 1-tert-butoxycarbonyl-3-hydroxy piperidine, boc-3-hydroxypiperidine, n-tert-butoxycarbonyl-3-piperidinol, n-boc-3-hydroxy piperidine, 3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 545699
IUPAC Name tert-butyl 3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

Melting Point 68°C
Color White
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-hydroxypiperidine