missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step Human Coupled IVT Kit - DNA

Catalog No. PI88881
Change view
Click to view available options
Quantity:
40 Reaction Kit
8 Reaction Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88881 8 Reaction Kit
PI88882 40 Reaction Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI88881 Supplier Thermo Scientific™ Supplier No. LSG88881

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rapidly express full-length, functional proteins from mRNA or plasmid templates with yields of up to 100μg/mL per reaction using these cell-free kits.

The Thermo Scientific™ 1-Step Human In Vitro Protein Expression Kits enable the translation and post-translational modification of full-length proteins from mRNA or plasmid templates with yields of up to 100 μg/mL per reaction.

The Human IVT Kits are unique HeLa cell lysate-based protein expression systems for in vitro translation or coupled transcription/translation reactions. Protein expression is performed in a single 90-minute reaction that can be extended for up to 6 hours with continued protein production up to 100μg/mL when combined with the optimized pT7CFE1 Expression Vector. The Human IVT Kits can express functional enzymes, phosphoproteins, glycoproteins and membrane proteins for immediate use in studying protein interactions, performing rapid mutational analysis and measuring activity.

Highlights:

Functional – uses the human translational machinery to express active proteins

  • Convenient – perform transcription and translation in a single step
  • High performance – greater yields compared to rabbit reticulocyte in vitro translation
  • Reliable – express proteins that fail in rabbit reticulocyte systems

Better Than Traditional Methods:

  • HeLa cell-free extracts are capable of expressing proteins with post-translational modifications
  • Accurate translation delivers full-length protein compatible with downstream applications
  • Protein translation is optimized with EMCV IRES element and other mRNA stabilizing elements

Includes:

Complete kits contain HeLa cell lysate, accessory proteins, reaction mix, nuclease-free water, expression vector and a GFP-positive control vector

Requires:

30°C incubator or water bath

Recommended for:

Express proteins to measure enzyme activity; Perform mutational analysis; Express protein for use in gel mobility shift assays; Perform co-immunoprecipitation; Express cytotoxic proteins; Perform unnatural amino acid labeling

Specifications

Type 1-step Human Coupled IVT Kit
Sufficient For 8 Reactions of 25 μL Each
Quantity 8 Reaction Kit
Content And Storage Sufficient For: 8 reactions of 25 μL each
• HeLa Lysate, 110 μL
• Accessory Proteins, 25 μL
• Reaction Mix, 40 μL
• Positive Control DNA: pCFE-GFP, 10 μg
• pT7CFE1-CHis, 10 μg
• Nuclease-Free Water, 1.5 mL

Upon receipt store kit at -80°C. Product is shipped on dry ice
Expression System Cell-Free
Expressed Protein Type Membrane Protein
Product Line 1-Step
What lysate systems can be used for in vitro protein expression?

E. coli, rabbit reticulocyte lysate (RRL), and HeLa cell lysate can all be used for in vitro translation. Our Expressway system utilizes E. coli. In general, RRL efficiently translates proteins greater than 30 kDa. The 1-Step Human In Vitro Protein Expression Kits are ideal for expressing human proteins.

Can I use 1-Step Human Coupled IVT Kit - DNA (Cat. No. 88881, 88882) with mRNA?

This page outlines a protocols for in vitro protein expression using mRNA templates.

When comparing 1-Step Human High-Yield Mini IVT Kit with 1-Step Human Coupled IVT Kit - DNA, is one faster than the other?

No, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA can be run overnight (depends upon the gene).

Do both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein?

Yes, both 1-Step Human High-Yield Mini IVT Kit and 1-Step Human Coupled IVT Kit - DNA provide the same purity of protein.

For Research Use Only. Not for use in diagnostic procedures.