missing translation for 'onlineSavingsMsg'
Learn More

1-acide pyrénéboronique (contient des quantités variables d’anhydride), TCI America™

Catalog No. P16251G
Change view
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
P16251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16251G Supplier TCI America Supplier No. P16251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 164461-18-1
Formule moléculaire C16H11BO2
Poids moléculaire (g/mol) 246.07
Numéro MDL MFCD04974062
Clé InChI MWEKPLLMFXIZOC-UHFFFAOYSA-N
Synonyme 1-pyrenylboronic acid, 1-pyreneboronic acid, pyrene-1-boronic acid, 1-boronopyrene, 1-pyrenyl boronic acid, boronic acid, 1-pyrenyl, 1-pyrene boronic acid, pyreneboronic acid, pubchem16490, pyren-1-yl boronic acid
PubChem CID 5084102
Nom de l’IUPAC (pyren-1-yl)acide boronique
SOURIRES OB(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Specifications

Nom ou substance chimique 1-Pyreneboronic Acid (contains varying amounts of Anhydride)
Point de fusion 251 °C
Quantité 1 g
Poids de formule 246.07
Forme physique Poudre cristalline