missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanéthiol 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -113°C |
| Color | Incolore |
| Boiling Point | 67°C |
| Quantity | 25 mL |
| UN Number | 2402 |
| Formula Weight | 76.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 1-Propanethiol |
Sécurité et manipulation
| EINECSNumber | (2)-0464 |
| RTECSNumber | TZ7300000 |
| TSCA | Yes |