missing translation for 'onlineSavingsMsg'
Learn More

1-Propanesulfonic acid, 95%

Catalog No. AC418340250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418340250 25 g Glass bottle
AC418340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC418340250 Supplier Thermo Scientific Chemicals Supplier No. 418340250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5284-66-2 | C3H8O3S | 124.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5284-66-2
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.16
MDL Number MFCD00041889
InChI Key KCXFHTAICRTXLI-UHFFFAOYSA-N
Synonym 1-propanesulfonic acid, propanesulfonic acid, propylsulfonic acid, propanesulfonate, propanesulphonic acid, ammonium propanesulfonate, propane-1-sulphonic acid, ncistruc1_001874, ncistruc2_000257, 1-propanesulfonic acid, technical t
PubChem CID 78938
IUPAC Name propane-1-sulfonic acid
SMILES CCCS(=O)(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 8.0°C
Color Brown
Density 1.2500g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Refractive Index 1.4380 to 1.4410
Linear Formula CH3CH2CH2SO3H
Quantity 25 g
Specific Gravity 1.25
Formula Weight 124.16
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Propanesulfonic acid, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 226-114-2
TSCA TSCA

RUO – Research Use Only