missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpyrrole 98.0+%, TCI America™

Catalog No. P11225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11225G 5 g
P112225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11225G Supplier TCI America Supplier No. P11225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 635-90-5
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00005343
InChI Key GEZGAZKEOUKLBR-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrrole, n-phenylpyrrole, pyrrole, 1-phenyl, 1h-pyrrole, 1-phenyl, unii-3od6n6545h, chembl84459, 1-phenylpyrrol, acmc-209ngl, 1-phenylpyrrole, 1-phenyl-1h-pyrrole #
PubChem CID 12480
IUPAC Name 1-phenyl-1H-pyrrole
SMILES C1=CN(C=C1)C1=CC=CC=C1

Specifications

Melting Point 59°C
Boiling Point 131°C
Quantity 5 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenylpyrrole