missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethyl Alcohol 98.0+%, TCI America™

Catalog No. M016325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M016325ML 25 mL
M0163500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M016325ML Supplier TCI America Supplier No. M016325ML

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004508
InChI Key WAPNOHKVXSQRPX-UHFFFAOYSA-N
Synonym methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
PubChem CID 7409
ChEBI CHEBI:669
IUPAC Name 1-phenylethanol
SMILES CC(C1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 mL
UN Number 2937
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (+/-)-1-Phenylethyl Alcohol
EINECSNumber (3)-1012&(3)-1049
RTECSNumber DO9275000
TSCA Yes