missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-Phenylethanol, 97%

Catalog No. AAA1383736
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1383736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1383736 Supplier Thermo Scientific Chemicals Supplier No. A1383736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 98-85-1 | C8H10O | 122.167 g/mol

The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The Arthrobacter sp. is able to grow with (+ I-,(-)-1-phenylethanol or the racemic mixture as sole source of carbon. Growth is most rapid with the (-)-isomer, doubling time 12 h. Lipases show good activity and, in some cases, improved enantioselectivity when employed in pure ionic liquids for dynamic kinetic resolution of 1-phenylethanol by transesterification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.

Chemical Identifiers

CAS 98-85-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004508
InChI Key WAPNOHKVXSQRPX-UHFFFAOYSA-N
Synonym methylphenyl carbinol, 1-phenylethan-1-ol, alpha-methylbenzyl alcohol, styralyl alcohol, 1-phenethyl alcohol, styrallyl alcohol, methylphenylcarbinol, 1-phenylethyl alcohol, phenylmethylcarbinol, 1-hydroxyethyl benzene
PubChem CID 7409
ChEBI CHEBI:669
IUPAC Name 1-phenylethanol
SMILES CC(C1=CC=CC=C1)O

Specifications

Melting Point 19°C to 20°C
Density 1.012
Boiling Point 203°C to 205°C
Quantity 500 g
UN Number UN2937
Solubility Information Insoluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material (+/-)-1-Phenylethanol
Hazard Category H227-H302-H319
Hazard Statement GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID
EINECSNumber 202-707-1
RTECSNumber DO9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only