missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-3-buten-1-ol 97.0+%, TCI America™

Catalog No. P13295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13295G Supplier TCI America Supplier No. P13295G

Chemical Identifiers

CAS 936-58-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00039617
InChI Key RGKVZBXSJFAZRE-UHFFFAOYSA-N
Synonym 1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
PubChem CID 220119
IUPAC Name 1-phenylbut-3-en-1-ol
SMILES C=CCC(C1=CC=CC=C1)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-3-buten-1-ol