missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-pyrrolidinone, 99%

Catalog No. AAL1466022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1466022 100 g
AAL1466006 5 g
AAL1466014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1466022 Supplier Thermo Scientific Chemicals Supplier No. L1466022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4641-57-0 | C10H11NO | 161.20 g/mol

A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

Solubility
Soluble in chloroform and ethyl Acetate. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 4641-57-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00003192
InChI Key JMVIVASFFKKFQK-UHFFFAOYSA-N
Synonym 1-phenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-phenyl, n-phenyl-2-pyrrolidone, n-phenyl-2-pyrrolidinone, 1-phenyl-pyrrolidin-2-one, 1-phenyl-2-pyrrolidone, n-phenylpyrrolidinone, n-phenyl pyrrolidone, pubchem23691, maybridge3_004960
PubChem CID 78375
IUPAC Name 1-phenylpyrrolidin-2-one
SMILES O=C1CCCN1C1=CC=CC=C1

Specifications

Melting Point 63°C to 68°C
Boiling Point 123°C (0.2 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 130685
Solubility Information Soluble in chloroform and ethyl Acetate. Insoluble in water.
Formula Weight 161.2
Percent Purity 99%
Chemical Name or Material 1-Phenyl-2-pyrrolidinone
EINECSNumber 225-069-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only