missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-propyn-1-one, 97%

Catalog No. AC456430010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456430010 Supplier Thermo Scientific Chemicals Supplier No. 456430010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3623-15-2 | C9H6O | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3623-15-2
Molecular Formula C9H6O
Molecular Weight (g/mol) 130.15
MDL Number MFCD06661646
InChI Key JITPLZPWKYUTDM-UHFFFAOYSA-N
Synonym 1-phenyl-2-propyn-1-one, propiolophenone, benzoylacetylene, propynophenone, propioloylbenzene, ethyne, benzoyl, ethynyl phenyl ketone, phenyl ethynyl ketone, 3-oxo-3-phenylpropyne, 2-propyn-1-one, 1-phenyl
PubChem CID 77179
IUPAC Name 1-phenylprop-2-yn-1-one
SMILES O=C(C#C)C1=CC=CC=C1

Specifications

Melting Point 51.0°C to 52.0°C
Boiling Point 90.0°C (0.8 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 130.15
Percent Purity 97%
Chemical Name or Material 1-Phenyl-2-propyn-1-one
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

RUO – Research Use Only