missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole 98.0+%, TCI America™

Catalog No. P22635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22635G 5 g
P22631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22635G Supplier TCI America Supplier No. P22635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 952514-86-2
Molecular Formula C25H25BN2O2
Molecular Weight (g/mol) 396.297
MDL Number MFCD16038225
InChI Key IFFQSTWSRPRHBQ-UHFFFAOYSA-N
Synonym 3-(1-Phenyl-1H-benzimidazole-2-yl)phenylboronic Acid Pinacol Ester
PubChem CID 58184542
IUPAC Name 1-phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzimidazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C3=NC4=CC=CC=C4N3C5=CC=CC=C5

Specifications

Melting Point 174°C
Color White
Quantity 5 g
Formula Weight 396.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-2-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-benzimidazole