missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%

Catalog No. AAH63695MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63695MD 250 mg
AAH6369503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63695MD Supplier Thermo Scientific Chemicals Supplier No. H63695MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54605-72-0 | C10H8N2O | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54605-72-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD02179568
InChI Key PHVRLPFVPVKYOI-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
PubChem CID 952089
IUPAC Name 1-phenylpyrazole-4-carbaldehyde
SMILES O=CC1=CN(N=C1)C1=CC=CC=C1

Specifications

Melting Point 84°C to 88°C
Color Yellow
Quantity 250 mg
Sensitivity Light and moisture sensitive
Formula Weight 172.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Phenyl-1H-pyrazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only