missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-butanol 98.0+%, TCI America™

Catalog No. P16825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16825G 5 g
P168225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16825G Supplier TCI America Supplier No. P16825G

Chemical Identifiers

CAS 614-14-2
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00066173
InChI Key HQRWWHIETAKIMO-UHFFFAOYSA-N
Synonym alpha-Propylbenzyl Alcohol
PubChem CID 95372
IUPAC Name 1-phenylbutan-1-ol
SMILES CCCC(C1=CC=CC=C1)O

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 170°C
Quantity 5 g
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-butanol
EINECSNumber (3)-1034
TSCA No