missing translation for 'onlineSavingsMsg'
Learn More

1-Phenacylpyridinium Bromide 98.0+%, TCI America™

Catalog No. P07975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07975G 5 g
P079725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07975G Supplier TCI America Supplier No. P07975G
Only null left

Chemical Identifiers

CAS 16883-69-5
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.149
MDL Number MFCD00031995
InChI Key PXSUMUYPXZEXDT-UHFFFAOYSA-M
Synonym 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PubChem CID 2798954
IUPAC Name 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
SMILES C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]

Specifications

Melting Point 198°C
Color White
Quantity 5 g
Formula Weight 278.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenacylpyridinium Bromide