missing translation for 'onlineSavingsMsg'
Learn More

1-Pentyne 98.0+%, TCI America™

Catalog No. P00685ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00685ML 5 mL
P006825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00685ML Supplier TCI America Supplier No. P00685ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-19-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.12
MDL Number MFCD00009469
InChI Key IBXNCJKFFQIKKY-UHFFFAOYSA-N
Synonym 1-pentyne, propylacetylene, pentyne, acetylene, propyl, pentin, pentyn, 1-n-pentyne, pent-1-yn-1-yl, acmc-1cuk3
PubChem CID 12309
IUPAC Name pent-1-yne
SMILES CCCC#C

Specifications

Color Yellow
Boiling Point 40°C
Quantity 5 mL
UN Number 3295
Formula Weight 68.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentyne