missing translation for 'onlineSavingsMsg'
Learn More

1-Penten-3-ol 97.0+%, TCI America™

Catalog No. P10435ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10435ML 5 mL
P104325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10435ML Supplier TCI America Supplier No. P10435ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

Color Colorless
Boiling Point 37°C
Quantity 5 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Penten-3-ol
EINECSNumber (2)-2393
TSCA Yes