missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Tolyl)piperazine 98.0+%, TCI America™
Supplier: TCI America T295625G
Specifications
| 1-(p-Tolyl)piperazine | |
| 36°C | |
| 112°C | |
| MFCD00040737 | |
| 3263 | |
| ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
| 1-(4-methylphenyl)piperazine | |
| 83113 | |
| ≥98.0% (GC,T) |
| 39593-08-3 | |
| White-Yellow | |
| C11H16N2 | |
| 25 g | |
| 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
| CC1=CC=C(C=C1)N2CCNCC2 | |
| 176.263 | |
| 176.26 | |
| Crystalline Lumps |
Chemical Identifiers
| 39593-08-3 | |
| 176.263 | |
| ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
| 83113 | |
| CC1=CC=C(C=C1)N2CCNCC2 |
| C11H16N2 | |
| MFCD00040737 | |
| 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
| 1-(4-methylphenyl)piperazine |
Safety and Handling
TSCA : No