missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(p-Tolyl)piperazine 98.0+%, TCI America™
$115.57 - $452.97
Chemical Identifiers
| CAS | 39593-08-3 |
|---|---|
| Molecular Formula | C11H16N2 |
| Molecular Weight (g/mol) | 176.263 |
| MDL Number | MFCD00040737 |
| InChI Key | ONEYFZXGNFNRJH-UHFFFAOYSA-N |
| Synonym | 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine |
| PubChem CID | 83113 |
| IUPAC Name | 1-(4-methylphenyl)piperazine |
| SMILES | CC1=CC=C(C=C1)N2CCNCC2 |
Chemical Identifiers
| 39593-08-3 | |
| 176.263 | |
| ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
| 83113 | |
| CC1=CC=C(C=C1)N2CCNCC2 |
| C11H16N2 | |
| MFCD00040737 | |
| 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
| 1-(4-methylphenyl)piperazine |
Specifications
| 39593-08-3 | |
| White-Yellow | |
| C11H16N2 | |
| 5 g | |
| 1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
| CC1=CC=C(C=C1)N2CCNCC2 | |
| 176.263 | |
| 176.26 | |
| Crystalline Lumps |
| 36°C | |
| 112°C | |
| MFCD00040737 | |
| 3263 | |
| ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
| 1-(4-methylphenyl)piperazine | |
| 83113 | |
| ≥98.0% (GC,T) | |
| 1-(p-Tolyl)piperazine |
Safety and Handling
TSCA : No