accessibility menu, dialog, popup

UserName

CAS RN 39593-08-3

IUPAC Name: 1-(4-methylphenyl)piperazine
Synonyms: 1-(4-methylphenyl)piperazine
Molecular Weight (g/mol): 176.26
Molecular Formula: C11H16N2
InChi Key: ONEYFZXGNFNRJH-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N1CCNCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)