missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog No. AAL1874606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1874606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1874606 Supplier Thermo Scientific Chemicals Supplier No. L1874606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3158-26-7 | Molecular Formula: C9H17NO | Molecular Weight: 155.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3158-26-7
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.241
MDL Number MFCD00013887
InChI Key DYQFCTCUULUMTQ-UHFFFAOYSA-N
Synonym octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
PubChem CID 76616
IUPAC Name 1-isocyanatooctane
SMILES CCCCCCCCN=C=O

Specifications

Density 0.88
Boiling Point 200°C to 204°C
Flash Point 71°C (159°F)
Odor Pungent
Refractive Index 1.432
Quantity 5 g
UN Number UN2206
Sensitivity Moisture sensitive
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-Octyl isocyanate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 221-598-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only