missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, Reagents

Numéro de catalogue. p-200020905
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01337151 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01337151 Fournisseur Labchem Inc Code fournisseur C2257850100C2
Il en reste null

1-Octanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym n-Octanol, n-Octyl Alcohol, normal Octanol
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Spécifications

Boiling Range 196°C
Melting Point -18°C
Color Colorless
Boiling Point 196°C
Melting Range -18°C
Packaging Glass Bottle
Quantity 100 g
Specific Gravity 0.827
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1-Octanol
Afficher plus de résultats Afficher moins de résultats