missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic Acid, Sodium Salt OR, Macron Fine Chemicals™

Catalog No. MKH16550
Change view
Click to view available options
Quantity:
50g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MKH16550 50g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MKH16550 Supplier Avantor Supplier No. H16556

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.271
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium;octane-1-sulfonate
SMILES CCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Packaging Glass
Quantity 50g
Formula Weight 234.28
Chemical Name or Material 1-Octanesulfonic Acid, Sodium Salt, Monohydrate