missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecylamine, 95%

Catalog No. AAL1545822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1545822 100 g
AAL1545814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1545822 Supplier Thermo Scientific Chemicals Supplier No. L1545822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 124-30-1 | C18H39N | 269.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1-Octadecylamine is used as asphalt emulsifier, lubricant and dispersants. It acts as a cationic surfactant and find applications is hair rinse bases, wood preservatives, textile softeners, dyeing auxiliaries and pigment grinding aids. It is used in boiler as an additive, which prevents the corrosion arises due to steam. It is a basic building block and used as an intermediate in the preparation of their amine derivatives, ethoxylates and amides. Metal oxide nano crystals having controlled size and shape are prepared by the thermal decomposition of metal nitrates with octadecylamine.

Solubility
Soluble in chloroform, alcohol, ether, benzene and acetone.Insoluble in water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 124-30-1
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.52
MDL Number MFCD00008159
InChI Key REYJJPSVUYRZGE-UHFFFAOYSA-N
Synonym octadecylamine, 1-octadecanamine, stearylamine, 1-octadecylamine, octadecanamine, stearamine, 1-aminooctadecane, n-octadecylamine, n-stearylamine, armofilm
PubChem CID 15793
ChEBI CHEBI:63866
IUPAC Name octadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCN

Specifications

Melting Point 53°C to 55°C
Density 0.862
Boiling Point 180°C to 183°C (5 mmHg)
Flash Point 148°C (298°F)
Linear Formula CH3(CH2)17NH2
Quantity 100 g
Beilstein 636111
Solubility Information Soluble in chloroform,alcohol,ether,benzene and acetone.Insoluble in water.
Formula Weight 269.52
Percent Purity 95%
Chemical Name or Material 1-Octadecylamine
Show More Show Less
Hazard Category H302-H314-H318-H373
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P262-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P310-P314-P330-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 204-695-3
RTECSNumber RG4150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only