missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanethiol 97.0+%, TCI America™

Catalog No. O000525G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O000525G 25 g
O0005400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O000525G Supplier TCI America Supplier No. O000525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2885-00-9
Molecular Formula C18H38S
Molecular Weight (g/mol) 286.56
MDL Number MFCD00004886
InChI Key QJAOYSPHSNGHNC-UHFFFAOYSA-N
Synonym octadecanethiol, 1-octadecanethiol, stearyl mercaptan, n-octadecyl mercaptan, octadecyl mercaptan, 1-mercaptooctadecane, octadecanthiol, n-octadecanethiol, octadecanethol, stearylmercaptan
PubChem CID 17905
IUPAC Name octadecane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCS

Specifications

Color White
Quantity 25 g
Formula Weight 286.56
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Octadecanethiol
EINECSNumber (2)-0464
TSCA Yes