missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-O-n-Octyl-beta-D-glucopyranoside, 98%
CAS: 29836-26-8 | C14H28O6 | 292.37 g/mol
$2,392.66
Chemical Identifiers
| CAS | 29836-26-8 |
|---|---|
| Molecular Formula | C14H28O6 |
| Molecular Weight (g/mol) | 292.37 |
| MDL Number | MFCD00063288 |
| InChI Key | HEGSGKPQLMEBJL-RKQHYHRCSA-N |
| Synonym | octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside |
| PubChem CID | 62852 |
| IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol |
| SMILES | CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC230340250
|
N/A | 25 g | Glass bottle |
Each for $2,392.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 29836-26-8 | |
| 292.37 | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| 62852 | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O |
| C14H28O6 | |
| MFCD00063288 | |
| octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol |
Specifications
| 29836-26-8 | |
| 100.0 | |
| White | |
| 97.5% min. (ELSD) (HPLC) | |
| MFCD00063288 | |
| − 32.00 (20.00°C c=5,water) | |
| octyl-beta-d-glucopyranoside, b-octylglucoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, unii-v109wut6rl, n-octyl glucoside | |
| HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
| (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol | |
| 62852 | |
| 98% | |
| 1-O-n-Octyl-β-D-glucopyranoside |
| 97.5 | |
| 105°C | |
| Authentic | |
| C14H28O6 | |
| 15, 6857 | |
| -32 | |
| (10% in water) Clear to slightly hazy colorless to light yellow | |
| CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O | |
| 292.37 | |
| 292.36 | |
| Crystalline Powder and/or Chunks |
Safety and Handling
EINECSNumber : 249-887-8
RUO – Research Use Only