missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2,5-bis(trifluorométhyl)benzène 98,0+%, TCI America™

Catalog No. B25465G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
B25465G 5 g
B254625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B25465G Supplier TCI America Supplier No. B25465G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 320-88-7
Formule moléculaire C8H3F6NO2
Poids moléculaire (g/mol) 259.107
Numéro MDL MFCD00236674
Clé InChI BZKVUOHHNCMTLH-UHFFFAOYSA-N
Synonyme 2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
PubChem CID 2734048
Nom de l’IUPAC 2-nitro-1,4-bis(trifluorométhyl)benzène
SOURIRES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F

Specifications

Nom ou substance chimique 1-Nitro-2,5-bis(trifluoromethyl)benzene
Point d’ébullition 90 °C
Quantité 5 g
Poids de formule 259.11
Pourcentage de pureté ≥98,0% (GC)
Forme physique Liquide