missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I01905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01905G 5 g
I019025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01905G Supplier TCI America Supplier No. I01905G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 551-06-4
Molecular Formula C11H7NS
Molecular Weight (g/mol) 185.244
MDL Number MFCD00003882
InChI Key JBDOSUUXMYMWQH-UHFFFAOYSA-N
Synonym 1-naphthyl isothiocyanate, 1-naphthylisothiocyanate, anit, kesscocide, alpha-naphthyl isothiocyanate, naphthalene, 1-isothiocyanato, naphthalene, isothiocyanato, isothiocyanic acid, 1-naphthyl ester, 1-naftylisothiokyanat, isothiocyanic acid 1-naphthyl ester
PubChem CID 11080
ChEBI CHEBI:35455
IUPAC Name 1-isothiocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=S

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 185.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthyl Isothiocyanate
RTECSNumber NX9100000
TSCA Yes