missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthoyl chloride, 99%

Catalog No. AC270380500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270380500 50 g Glass bottle
AC270380100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270380500 Supplier Thermo Scientific Chemicals Supplier No. 270380500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 879-18-5 | C11H7ClO | 190.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 879-18-5
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.63
MDL Number MFCD00004002
InChI Key NSNPSJGHTQIXDO-UHFFFAOYSA-N
Synonym 1-naphthoyl chloride, 1-naphthalenecarbonyl chloride, 1-naphthoylchloride, 1-naphthoic acid chloride, 1-chlorocarbonyl naphthalene, alpha-naphthoyl chloride, .alpha.-naphthoyl chloride, naphthalenecarbonyl chloride, naphthoylchloride, naphthoyl chloride
PubChem CID 70146
IUPAC Name naphthalene-1-carbonyl chloride
SMILES ClC(=O)C1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 16°C to 19°C
Color White
Density 1.2600g/mL
Boiling Point 190°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6515 to 1.6535
Quantity 50 g
Beilstein 09, 648
Specific Gravity 1.26
Solubility Information Solubility in water: decomposes
Formula Weight 190.63
Percent Purity 99%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material 1-Naphthoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 212-903-9

RUO – Research Use Only