missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. N0017500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0017500G 500 g
N001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0017500G Supplier TCI America Supplier No. N0017500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-46-1
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.674
MDL Number MFCD00003984
InChI Key DASJFYAPNPUBGG-UHFFFAOYSA-N
Synonym 1-naphthalenesulfonyl chloride, 1-naphthylsulfonyl chloride, naphthalene-1-sulphonyl chloride, alpha-naphthalenesulfonyl chloride, naphthalenesulfonyl chloride, 1-naphthalenesulfonylchloride, 1-naphthalenesulfonyl chlorine, .alpha.-naphthalenesulfochloride, chloronaphthylsulfone, .alpha.-naphthalenesulfonyl chloride
PubChem CID 66560
IUPAC Name naphthalene-1-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)Cl

Specifications

Melting Point 70°C
Color White
Boiling Point 195°C
Quantity 500 g
UN Number 3261
Formula Weight 226.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Naphthalenesulfonyl Chloride
EINECSNumber (4)-1096
TSCA Yes